Learn More
Tirbanibulin Mesylate, MedChemExpress
MedChemExpress Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.
Brand:  Medchem Express HY-10340B
Encompass_Preferred
 View more versions of this product
Product Code. 16430217
  • 4979.39 SEK / 50mg
Explore more special offers
Description
Description
Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.
Specifications
Specifications
Tirbanibulin Mesylate
Off-White
C27H33N3O6S
KX2-391 MesylateKX01 Mesylate
O=C(NCC1=CC=CC=C1)CC2=NC=C(C3=CC=C(OCCN4CCOCC4)C=C3)C=C2.CS(=O)(O)=O
527.63
Research
Cancer-Kinase/protease
1080645-95-9
Research
50 mg
DMSO : 100 mg/mL (189.53 mM; Need ultrasonic)
527.63
99.97%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Tirbanibulin Mesylate, MedChemExpress