Learn More
Tropisetron Hydrochloride, MedChemExpress
MedChemExpress Tropisetron Hydrochloride (SDZ-ICS-930) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.
Brand:  Medchem Express HY-B0020
Encompass_Preferred
 View more versions of this product
Product Code. 16418328
  • 1058.12 SEK / 10mg
Explore more special offers
Description
Description
Tropisetron Hydrochloride (SDZ-ICS-930) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.
Specifications
Specifications
Tropisetron Hydrochloride
105826-92-4
10 mg
C17H21ClN2O2
SDZ-ICS-930
O=C(C1=CNC2=C1C=CC=C2)O[C@H]3C[C@H]4CC[C@H](N4C)C3.[H]Cl
320.81
Research
Solid
White
Research
Neuroscience-Neuromodulation
H2O : ≥ 50 mg/mL (155.86 mM) ∣DMSO : 33.33 mg/mL (103.89 mM; Need ultrasonic)
320.81
99.95%
Documents
One moment while we fetch your results.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Tropisetron Hydrochloride, MedChemExpress