Learn More
Vistusertib, MedChemExpress
MedChemExpress Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.
Brand:  Medchem Express HY-15247
Encompass_Preferred
 View more versions of this product
Product Code. 16421176
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.
Specifications
Specifications
Vistusertib
Yellow
C25H30N6O3
AZD2014
O=C(NC)C1=CC=CC(C2=CC=C3C(N=C(N4[C@@H](C)COCC4)N=C3N5[C@@H](C)COCC5)=N2)=C1
462.54
Research
Cancer-Kinase/protease
1009298-59-2
Research
200 mg
DMSO : ≥ 50 mg/mL (108.10 mM)
462.54
95.66%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Vistusertib, MedChemExpress