Learn More
Applied Biosystems™ Anode Buffer Container (ABC), for 3500/SeqStudio™ Flex
GREENER_CHOICE
The Anode Buffer Container (ABC) contains 1X running buffer to support all electrophoresis applications on the Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.
Brand:  Applied Biosystems™ 4393927
1296.75 SEK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15809722
  • 1365.00 SEK / Pack of 4
Explore more special offers
Description
Description
The Anode Buffer Container (ABC) contains 1X running buffer to support all electrophoresis applications on the Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers. It is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the ABC is heat sealed with a plastic film, which should be removed prior to direct installation on the instrument. Each package includes four individual containers.
Order Info
4 Pack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
4 pack
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2411984 Certificate TypeCertificate of Analysis Date24/01/2025 Catalog Number4393927
Lot Number2411983 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number4393927
Lot Number2411982 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog Number4393927
Lot Number2410981 Certificate TypeCertificate of Analysis Date08/01/2025 Catalog Number4393927
Lot Number2410980 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number4393927
Product Title
Applied Biosystems™ Anode Buffer Container (ABC), for 3500/SeqStudio™ Flex >