Learn More
Lacidipine, MedChemExpress
MedChemExpress Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Brand:  Medchem Express HY-B0347
Encompass_Preferred
 View more versions of this product
Product Code. 16491406
  • POA / 200mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Specifications
Specifications
Lacidipine
Off-White
C26H33NO6
DMSO : ≥ 50 mg/mL (109.76 mM)
455.54
99.98%
Solid
103890-78-4
Research
200 mg
O=C(C1=C(C)NC(C)=C(C(OCC)=O)C1C2=CC=CC=C2/C=C/C(OC(C)(C)C)=O)OCC
455.54
Research
COVID-19-immunoregulation
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Lacidipine, MedChemExpress