Learn More
Lacidipine, MedChemExpress
MedChemExpress Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Brand: Medchem Express HY-B0347
Description
Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.Specifications
Lacidipine | |
Off-White | |
C26H33NO6 | |
DMSO : ≥ 50 mg/mL (109.76 mM) | |
455.54 | |
99.98% | |
Solid |
103890-78-4 | |
Research | |
200 mg | |
O=C(C1=C(C)NC(C)=C(C(OCC)=O)C1C2=CC=CC=C2/C=C/C(OC(C)(C)C)=O)OCC | |
455.54 | |
Research | |
COVID-19-immunoregulation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only